Specifications
- Antibody type:Primary
- Antigen name:Rhodopsin
- Antigen symbol:RHO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Tiger finch,Zebra finch,Rat,Mouse
- Western blot:Yes
- Epitope:Peptides cooresponding to amino acids 14-26 of Mouse Opsin 1 and amino acids 18-30 of Tiger finch opsin1.
- Cross adsorption:Yes
- Format:Liquid
- Immunogen:Mixture of synthetic peptide (14-26 aa) of Mouse Green cones Opsin 1 and synthetic peptide (18-30 aa) of Tiger finch red cones opsin 1 was used as immunogen.
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Rod shaped photoreceptor cells that are required for image-forming vision at low light intensity and for photoreceptor cell viability after birth (ref: SWISSPROT).
The antibody reacts in wester blots in mouse brain extracts with a prominent band ~36kDa cooresponding to Rhodopsin. The antibody has not been tested in IHC.
Application Information:
WB. Suggested dilution at 1:500 to 1:2,000. To minimise background staining, a higher concentration of detergent (such as Tween-20) is suggested in the dilution and washing steps. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Rhodopsin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Peptides cooresponding to amino acids 14-26 of Mouse Opsin 1 and amino acids 18-30 of Tiger finch opsin1.
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat, Tiger Finch, Zebra Finch