Specifications
- Antibody type:Primary
- Antigen name:Orexin B
- Antigen symbol:HCRT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Rat,Mouse
- Epitope:Human Orexin B (1-14 aa).
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:Hypocretin-2|Hcrt2|Orexin
- Immunogen:A peptide from the N-terminus of human Orexin B (1-14 aa).
- Size:100 µl
- Pk:100 µl
Specifications
About this item
FUNCTION: Neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity. SUBCELLULAR LOCATION: Endoplasmic reticulum; rough endoplasmic reticulum. Associated with perikaryal rough endoplasmic reticulum as well as cytoplasmic large granular vesicles at synapses. SIMILARITY: Belongs to the orexin family.
The antibody recognizes Orexin-B in ELISA other applications have not been tested.
Application Information:
ELISA . Suggested dilution at 1:2,000 to 1:5,000 as a detection antibody and 1:200 to 1:1,000 as a coating antibody in sandwich ELISA. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Orexin B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Orexin B (1-14 aa).
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat