Anti-SCNN1G Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # ABNOPAB28818
Supplier: Abnova
Specifications
- Antigen name:Sodium Channel Nonvoltage-gated 1 gamma
- Antigen symbol:SCNN1G
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:24768
- Antigen synonyms:Epithelial Na(+) channel subunit gamma|ENaCg|SCNEG)|PHA1|Amiloride-sensitive sodium channel subunit gamma (SCNN1G|Gamma-NaCH|SCNEG,|Gamma-ENaC
- Storage buffer:PBS, 50% glycerol and 0,09% sodium azide
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to Scnn1g C-terminus (amino acids 629-650).
- Purification:Affinity purification
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant Rat Scnn1g.
Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000
Type:
Antigen: SCNN1G
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype:
Reactivity: