Order Entry
Netherlands
ContactUsLinkComponent
Anti-HTR2C Rabbit Polyclonal Antibody
Anti-HTR2C Rabbit Polyclonal Antibody
Catalog # ABNOPAB25547
Supplier:  Abnova
Anti-HTR2C Rabbit Polyclonal Antibody
Catalog # ABNOPAB25547
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5-hydroxytryptamine (serotonin) receptor 2C
  • Antigen symbol:
    HTR2C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Chimpanzee,
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3358
  • Antigen synonyms:
    5-HT1C|5-HTR2C|5HTR2C|HTR1C|5-HT2C
  • Storage buffer:
    PBS (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 16 amino acids at C-terminus of human HTR2C.
  • Purification:
    Immunoaffinity chromatography
  • Size:
    50 μg
  • Pk:
    50 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of HTR2C.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 3.8 ?g/ml

Type: Primary
Antigen: HTR2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Human