Anti-MATK Mouse Monoclonal Antibody [clone: 9D7]
Catalog # ABNOMAB10803
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Megakaryocyte-associated Tyrosine Kinase
- Antigen symbol:MATK
- Clonality:Monoclonal
- Clone:9D7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4145
- Antigen synonyms:HYLTK|Tyrosine-protein kinase CTK|CTK|HHYLTK|Lsk|Protein kinase HYL|Megakaryocyte-associated tyrosine-protein kinase|CHK|Hematopoietic consensus tyrosine-lacking kinase|HYL
- Storage buffer:PBS (50% glycerol, 0,03% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human MATK.
- Purification:Protein G purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant MATK.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: MATK
Clonality: Monoclonal
Clone: 9D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human