Anti-ALPP Mouse Monoclonal Antibody [clone: NB-10]
Catalog # ABNOMAB10731
Leverancier: Abnova
Specificaties
- Antibody type:Primary
- Antigen name:Alkaline Phosphatase, Placental
- Antigen symbol:ALPP
- Clonality:Monoclonal
- Clone:NB-10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:250
- Antigen synonyms:PALP|ALP|PLAP|PLAP-1
- Storage buffer:PBS, pH 7,4 (protebase, 0,09% sodium azide)
- Storage temperature:Store at 4 °C is stable for 3 years.
- Immunogen:ALPP.
- Size:1 mL
- Pk:1 ml
Specificaties
Over dit artikel
Mouse monoclonal antibody raised against ALPP.
Type: Primary
Antigen: ALPP
Clonality: Monoclonal
Clone: NB-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human