Anti-COX4I1 Mouse Monoclonal Antibody [clone: 6B3]
Catalog # ABNOMAB10611
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome C Oxidase Subunit Iv Isoform 1
- Antigen symbol:COX4I1
- Clonality:Monoclonal
- Clone:6B3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:1327
- Antigen synonyms:COXIV|IV-1|AL024441|COX|COX IV-1|Cox4a|Cox4
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human COX4I1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant COX4I1.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: COX4I1
Clonality: Monoclonal
Clone: 6B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat