Anti-HTR2B Rabbit Polyclonal Antibody
ABNOH00003357-D01P
: Abnova
- Antibody type:Primary
- Antigen name:5-hydroxytryptamine (serotonin) receptor 2B
- Antigen symbol:HTR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3357
- Antigen synonyms:5-HT2B|5-HT(2B)
- Amino acid number:1 to 481
- Storage buffer:1x PBS, pH 7,4
- Sequence:MALSYRVSELQSTIPEHILQSTFVHVISSNWSGLQTESIPEEMKQIVEEQGNKLHWAALLILMVIIPTIGGNTLVILAVSLEKKLQYATNYFLMSLAVADLLVGLFVMPIALLTIMFEAMWPLPLVLCPAWLFLDVLFSTASIMHLCAISVDRYIAIKKPIQANQYNSRATAFIKITVVWLISIGIAIPVPIKGIETDVDNPNNITCVLTKERFGDFMLFGSLAAFFTPLAIMIVTYFLTIHALQKKAYLVKNKPPQRLTWLTVSTVFQRDETPCSSPEKVAMLDGSRKDKALPNSGDETLMRRTSTIGKKSVQTISNEQRASKVLGIVFFLFLLMWCPFFITNITLVLCDSCNQTTLQMLLEIFVWIGYVSSGVNPLVYTLFNKTFRDAFGRYITCNYRATKSVKTLRKRSSKIYFRNPMAENSKFFKKHGIRNGINPAMYQSPMRLRSSTIQSSSIILLDTLLLTENEGDKTEERVSYV
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:HTR2B (NP_000858.2, 1 a.a. ~ 481 a.a) full-length human protein.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against a full-length human HTR2B protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: HTR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human