Order Entry
Netherlands
ContactUsLinkComponent
Anti-HLA-E Mouse Monoclonal Antibody [clone: MEM-E/02]
Anti-HLA-E Mouse Monoclonal Antibody [clone: MEM-E/02]
Catalog # ABNOMAB0939
Supplier:  Abnova
Anti-HLA-E Mouse Monoclonal Antibody [clone: MEM-E/02]
Catalog # ABNOMAB0939
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA class I histocompatibility antigen, E alpha chain
  • Antigen symbol:
    HLA-E
  • Clonality:
    Monoclonal
  • Clone:
    MEM-E/02
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Primates,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3133
  • Antigen synonyms:
    EA2.1|HLA class I histocompatibility antigen|HLA-6.2|lymphocyte antigen|MHC HLA-E alpha-1|E alpha chain|DAMA-277I14.1|DKFZp686P19218|MHC class I antigen E|EA1.2|MHC|alpha chain E|QA1|MHC HLA-E alpha-2.1
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to heavy chain region (denaturated) of human HLA-E.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant HLA-E.

Type: Primary
Antigen: HLA-E
Clonality: Monoclonal
Clone: MEM-E/02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primates