Anti-CD3E Mouse Monoclonal Antibody [clone: MEM-92]
ABNOMAB0952
: Abnova
- Antibody type:Primary
- Antigen name:CD3 Molecule Epsilon
- Antigen symbol:CD3E
- Clonality:Monoclonal
- Clone:MEM-92
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:916
- Antigen synonyms:CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified CD3E from human peripheral blood lymphocytes.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native CD3E.
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: MEM-92
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human