Anti-MME Mouse Monoclonal Antibody [clone: MEM-78]
Catalog # ABNOMAB0875
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Membrane metallo-endopeptidase
- Antigen symbol:MME
- Clonality:Monoclonal
- Clone:MEM-78
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:4311
- Antigen synonyms:NEP|CD10|CALLA|SFE
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified MME from human pre-B cell line NALM-6.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native MME.
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: MEM-78
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human