Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD48 Mouse Monoclonal Antibody [clone: MEM-102]
Anti-CD48 Mouse Monoclonal Antibody [clone: MEM-102]
Catalog # ABNOMAB0889
Supplier:  Abnova
Anti-CD48 Mouse Monoclonal Antibody [clone: MEM-102]
Catalog # ABNOMAB0889
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD48 molecule
  • Antigen symbol:
    CD48
  • Clonality:
    Monoclonal
  • Clone:
    MEM-102
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Primates,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    962
  • Antigen synonyms:
    MEM-102|BLAST|hCD48|mCD48|BLAST1|BCM1|SLAMF2
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified CD48 from Raji human Burkitt's lymphoma cell line.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native CD48.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: MEM-102
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primates