Anti-HRP Monoclonal Antibody [clone: NI 91]
ABNOMAB10077
: Abnova
- Antibody type:Primary
- Antigen name:Horseradish Peroxidase
- Antigen symbol:HRP
- Clonality:Monoclonal
- Clone:NI 91
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:HRP|Horseradish Peroxidase
- Storage buffer:PBS, pH 7,2
- Storage temperature:Store at 4 °C on dry atmosphere. After reconstitution with 0.5 mL deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Purified HRP isomer 1.
- Size:200 μg
- Pk:200 µg
Mouse monoclonal antibody raised against HRP.
Recommended Dilutions: ELISA: 1:500; Western Blot: 1:1000; Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: HRP
Clonality: Monoclonal
Clone: NI 91
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: