Anti-SMAD4 Mouse Monoclonal Antibody [clone: IMD-89]
Catalog # ABNOMAB1485
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:SMAD family member 4
- Clonality:Monoclonal
- Clone:IMD-89
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:4089
- Antigen synonyms:Drosophila|mothers against decapentaplegic homolog 4,SMAD 4|hSMAD4|SMAD|mothers against decapentaplegic homolog 4|mothers against DPP homolog 4|MAD|deleted in pancreatic carcinoma locus 4|4|MAD homolog 4|mothers against decapentaplegic|deletion target in pancreatic carcinoma 4|homolog of
- Storage buffer:1.2% sodium acetate (2 mg BSA, 0,01 mg sodium azide)
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with 1 mL of 1.2% sodium acetate or neutral PBS and concentration will be 100 ug/mL, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human SMAD4.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant SMAD4.
Recommended Dilutions: Western Blot: 2-4 ?g/ml
Type: Primary
Antigen: SMAD4
Clonality: Monoclonal
Clone: IMD-89
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human