Anti-FCGR1C Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 10.1]
Catalog # ABNOMAB10245
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fc fragment of IgG, high affinity Ic, receptor (CD64), pseudogene
- Antigen symbol:FCGR1C
- Clonality:Monoclonal
- Clone:10.1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Primates,Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:100132417
- Antigen synonyms:IGFRC|FCRIC|CD64c|IGFR1
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Rheumatoid synovial fluid cells and fibronectin purified human monocytes.
- Size:100 Reactions
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against of FCGR1C.
Recommended Dilutions: Flow Cytometry: 4 ?l reagent/100 ?l of whole blood or 10^6 cells
Type: Primary
Antigen: FCGR1C
Clonality: Monoclonal
Clone: 10.1
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-human Primates