Order Entry
Netherlands
ContactUsLinkComponent
Anti-FCGR1C Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10.1]
Anti-FCGR1C Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10.1]
Catalog # ABNOMAB10246
Supplier:  Abnova
Anti-FCGR1C Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10.1]
Catalog # ABNOMAB10246
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fc fragment of IgG, high affinity Ic, receptor (CD64), pseudogene
  • Antigen symbol:
    FCGR1C
  • Clonality:
    Monoclonal
  • Clone:
    10.1
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Primates,
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    100132417
  • Antigen synonyms:
    IGFRC|FCRIC|CD64c|IGFR1
  • Storage buffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Immunogen:
    Rheumatoid synovial fluid cells and fibronectin purified human monocytes.
  • Purification:
    Size-exclusion chromatography purification
  • Size:
    100 Reactions
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against of FCGR1C.

Recommended Dilutions: Flow Cytometry: 10 ?l reagent/100 ?l of whole blood or 10^6 cells

Type: Primary
Antigen: FCGR1C
Clonality: Monoclonal
Clone: 10.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-human Primates