Anti-SRCAP Rabbit Polyclonal Antibody
Catalog # ABNOPAB22228
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Snf2-related CREBBP activator Protein
- Antigen symbol:SRCAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:10847
- Antigen synonyms:FLJ44499|KIAA0309|helicase SRCAP|domino homolog 1|EAF1|SWR1|domino homolog 2|Swi2/Snf2-related ATPase homolog domino homolog 1|Snf2-related CREBBP activator protein|DOMO1|Snf2-related CBP activator protein
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:AQALASPESLELASVASSETSSLSLVPPKDLLPVAVEILPVSEKNLSLTPSAPSLTLEAGSIPNGQEQEAPDSAEGTTLTVLPEGEELPLCVSESNGLELPPSAASDEPLQEPLEADRTSEELTEAKTPTSS
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human SRCAP.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against recombinant SRCAP.
Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Immunofluorescence: 1-4 ?g/ml
Type: Primary
Antigen: SRCAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human