Anti-SLC5A7 Mouse Monoclonal Antibody [clone: 62-2E8]
ABNOMAB2295
: Abnova
- Antibody type:Primary
- Antigen name:Solute Carrier Family 5 (Choline transporter) Member 7
- Antigen symbol:Solute carrier family 5, member 7
- Clonality:Monoclonal
- Clone:62-2E8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:60482
- Antigen synonyms:HMN7A|CHT|CHT1
- Storage buffer:PBS (0,05% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to C-terminus of human SLC5A7.
- Size:100 μl
- Pk:100 µg
Mouse monoclonal antibody raised against partial recombinant SLC5A7.
Recommended Dilutions: Western Blot: 1:500; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:500; Immunohistochemistry (Frozen sections): 1:500
Type: Primary
Antigen: SLC5A7
Clonality: Monoclonal
Clone: 62-2E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat