Order Entry
Netherlands
ContactUsLinkComponent
Anti-HBcAg Monoclonal Antibody [clone: C1]
Anti-HBcAg Monoclonal Antibody [clone: C1]
  ABNOMAB4775
 :  Abnova
Anti-HBcAg Monoclonal Antibody [clone: C1]
  ABNOMAB4775
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Hepatitis B Core Antigen
  • Antigen symbol:
    HBcAg
  • Clonality:
    Monoclonal
  • Clone:
    C1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Antigen synonyms:
    Hepatitis B virus nucleocapsid particles|HepB
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to hepatitis B core antigen.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against recombinant hepatitis B core Antigen.

Type: Primary
Antigen: HBcAg
Clonality: Monoclonal
Clone: C1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity: Virus