Anti-UCHL1 Mouse Monoclonal Antibody [clone: BH7]
Catalog # ABNOMAB2381
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen symbol:UCHL1
- Clonality:Monoclonal
- Clone:BH7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Tissue Culture Supernatant
- Gene ID:7345
- Antigen synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Storage buffer:10 mM sodium azide
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human UCHL1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant UCHL1.
Recommended Dilutions: Immunofluorescence: 1:1000; Western Blot: 1:10000
Type: Primary
Antigen: UCHL1
Clonality: Monoclonal
Clone: BH7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Rat