Anti-HPGDS Rabbit Polyclonal Antibody
ABNOPAB13619
: Abnova
- Antibody type:Primary
- Antigen name:Hematopoietic prostaglandin D synthase
- Antigen symbol:HPGDS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:58962
- Antigen synonyms:GSTS1-1|PGDS|GSTS1|PGD2|GSTS
- Storage buffer:buffer containing 0,05% sodium azide
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Rat Ptgds2.
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against Ptgds2.
Recommended Dilutions: Western Blot: 1:800; Immunohistochemistry: 1:800
Type: Primary
Antigen: HPGDS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat