Order Entry
Netherlands
ContactUsLinkComponent
Anti-HLA-DR+DP Monoclonal Antibody (Biotin) [clone: MEM-136]
Anti-HLA-DR+DP Monoclonal Antibody (Biotin) [clone: MEM-136]
Catalog # ABNOMAB5069
Supplier:  Abnova
Anti-HLA-DR+DP Monoclonal Antibody (Biotin) [clone: MEM-136]
Catalog # ABNOMAB5069
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HLA-DR+DP
  • Clonality:
    Monoclonal
  • Clone:
    MEM-136
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified from PHA-activated peripheral blood lymphocytes.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native HLA-DR+DP.

Recommended Dilutions: Flow Cytometry: 1:400

Type: Primary
Antigen: HLA-DR+DP
Clonality: Monoclonal
Clone: MEM-136
Conjugation: Biotin
Epitope:
Host:
Isotype: IgG1
Reactivity: Human