Anti-UCHL1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB11988
Leverancier: Abnova
Specificaties
- Antibody type:Primary
- Antigen name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen symbol:UCHL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Primates,Pig,Horse,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7345
- Antigen synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Amino acid number:200 to 300
- Storage buffer:Tris-glycine, 150 mM NaCl (0,05% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 200-300 of human UCHL1.
- Size:100 μl
- Pk:100 µg
Specificaties
Over dit artikel
Rabbit polyclonal antibody raised against synthetic peptide of UCHL1.
Recommended Dilutions: Western Blot: 2 ?g/ml; Immunohistochemistry: 2.5-10 ?g/ml
Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Horse, Human, Mouse, Pig, Primates, Rat