Order Entry
Netherlands
ContactUsLinkComponent
Anti-NFYA Rabbit Polyclonal Antibody
Anti-NFYA Rabbit Polyclonal Antibody
Catalog # ABNOPAB11297
Supplier:  Abnova
Anti-NFYA Rabbit Polyclonal Antibody
Catalog # ABNOPAB11297
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear transcription factor Y, alpha
  • Antigen symbol:
    NFYA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    4800
  • Antigen synonyms:
    CBF-A|CBF-B|NF-YA|HAP2
  • Storage buffer:
    20 mM KH2PO4, 150 mM NaCl, pH 7,2 (0,01% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human NFYA.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of NFYA.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000

Type: Primary
Antigen: NFYA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human