Anti-HLA-DPB1 Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEM-136]
ABNOMAB4484
: Abnova
- Antibody type:Primary
- Antigen symbol:HLA-DPB1
- Clonality:Monoclonal
- Clone:MEM-136
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3115|3123|3125|3126|3127
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native from PHA-activated peripheral blood leucocytes.
- Size:100 Reactions
- Pk:100 µl
Mouse monoclonal antibody raised against HLA-DR/HLA-DP.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: HLA-DPB1
Clonality: FITC (Fluorescein Isothiocyanate)
Clone: Monoclonal
Conjugation: MEM-136
Epitope:
Host: IgG1
Isotype:
Reactivity: Human