Anti-CFLAR Mouse Monoclonal Antibody [clone: 5D8]
Catalog # ABNOMAB2065
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CASP8 and FADD-like apoptosis regulator
- Antigen symbol:CFLAR
- Clonality:Monoclonal
- Clone:5D8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:8837
- Antigen synonyms:c-FLIP|c-FLIPS|CLARP|FLAME-1|I-FLICE|MRIT|CASP8AP1|FLAME|c-FLIPL|c-FLIPR|FLIP|FLAME1|CASH|Casper
- Amino acid number:1 to 376
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 1-376 of human CFLAR.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CFLAR.
Type: Primary
Antigen: CFLAR
Clonality: Monoclonal
Clone: 5D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human, Mouse, Rat