Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-61]
Anti-CD9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-61]
Catalog # ABNOMAB4559
Supplier:  Abnova
Anti-CD9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-61]
Catalog # ABNOMAB4559
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD9 molecule
  • Antigen symbol:
    CD9
  • Clonality:
    Monoclonal
  • Clone:
    MEM-61
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    928
  • Antigen synonyms:
    MIC3|TSPAN29|TSPAN-29|DRAP-27|MRP-1|BTCC-1
  • Storage buffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified CD9 from Pre-B cell line NALM-6.
  • Size:
    100 Reactions
  • Pk:
    100 Tests

Specifications

About this item

Mouse monoclonal antibody raised against native CD9.

Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: MEM-61
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human