Anti-MME Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-78]
Catalog # ABNOMAB4605
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Membrane metallo-endopeptidase
- Antigen symbol:MME
- Clonality:Monoclonal
- Clone:MEM-78
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:4311
- Antigen synonyms:NEP|CD10|CALLA|SFE
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified MME from NALM-6 human pre-B cell line.
- Size:100 Reactions
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native MME.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: MEM-78
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human