Anti-GRINA Rabbit Polyclonal Antibody
ABNOPAB23092
: Abnova
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1
- Antigen symbol:GRINA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:2907
- Antigen synonyms:TMBIM3|NMDA receptor glutamate-binding subunit|LFG1|NMDARA1|GRINA|HNRGW|Protein lifeguard 1
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:PYAQPPYPGAPYPQPPFQPSPYGQPGYPHGPSPYPQGGYPQGPY
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human GRINA.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant GRINA.
Recommended Dilutions: Immunohistochemistry: 1:20-1:50
Type: Primary
Antigen: GRINA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human