Anti-INSM2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB16839
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Insulinoma-associated 2
- Antigen symbol:INSM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:84684
- Antigen synonyms:IA6|IA-6|Zinc finger protein IA-6|Insulinoma-associated protein 2|Nbla106|mlt1
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 14 amino acids of human INSM2.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of INSM2.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: INSM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat