Anti-HLA-DQA1 Mouse Monoclonal Antibody [clone: TAL 4.1]
ABNOMAB7017
: Abnova
- Antibody type:Primary
- Antigen name:major histocompatibility complex, class II, DQ alpha 1
- Antigen symbol:HLA-DQA1
- Clonality:Monoclonal
- Clone:TAL 4.1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3117
- Antigen synonyms:HLA-DQA|GSE|CD|DQ-A1|CELIAC1
- Storage buffer:PBS
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified HLA-DQA1 from whole cells from the B line WT46.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native HLA-DQA1.
Type: Primary
Antigen: HLA-DQA1
Clonality: Monoclonal
Clone: TAL 4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human