Anti-TAAR5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB16376
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Trace amine associated receptor 5
- Antigen symbol:TAAR5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rabbit,Gorilla,Chimpanzee,Human,Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:9038
- Antigen synonyms:Putative neurotransmitter receptor|TAAR5|Trace amine associated receptor 5|Trace amine-associated receptor 5
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to human TAAR5.
- Purification:Immunoaffinity purification
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of TAAR5.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 20 ?g/ml
Type: Primary
Antigen: TAAR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Human, Mouse, Rabbit