Anti-UCHL3 Rabbit Polyclonal Antibody
ABNOPAB16472
: Abnova
- Antibody type:Primary
- Antigen name:Ubiquitin carboxyl-terminal esterase L3
- Antigen symbol:UCHL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Horse,Monkey,Gorilla,Orangutan,Human,Bovine,Dog,Rat,Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:7347
- Antigen synonyms:Ubiquitin thioesterase L3|Ubiquitin thiolesterase L3|UCH L3|UCH-L3|UCHL3_HUMAN.|Ubiquitin thiolesterase
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to human UCHL3.
- Purification:Immunoaffinity purification
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against synthetic peptide of UCHL3.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10 ?g/ml
Type: Primary
Antigen: UCHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Gorilla, Horse, Human, Monkey, Mouse, Orangutan, Rat