Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-MBD2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB14065
Supplier:  Abnova
Anti-MBD2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB14065
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Methyl-CpG binding domain protein 2
  • Antigen symbol:
    MBD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Serum
  • Gene ID:
    17191
  • Antigen synonyms:
    MBD2a|NY-CO-41|DMTase
  • Storage buffer:
    0,05% sodium azide
  • Storage temperature:
    Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide (conjugated with KLH) corresponding to internal region of mouse Mbd2.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of Mbd2.

Recommended Dilutions: ELISA: 1:1000-1:4000; Western Blot: 1:500

Type: Primary
Antigen: MBD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse