Anti-CEL Monoclonal Antibody (Biotin) [clone: KNH-30]
Catalogus # ABNOMAB6594
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Carboxyl ester lipase
- Antigen symbol:CEL
- Clonality:Monoclonal
- Clone:KNH-30
- Conjugation:Biotin
- ELISA:Yes
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:FAP|BSDL|FAPP|MODY8|LIPA|BAL|BSSL|CEase|CELL
- Storage buffer:2% Block Ace, 0,1% proclin
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein (conjugated with BSA) corresponding to CEL.
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant CEL.
Recommended Dilutions: ELISA: 0.1-1 ?g/ml; Immunohistochemistry: 5-10 ?g/ml
Type: Primary
Antigen: CEL
Clonality: Monoclonal
Clone: KNH-30
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse, Rat