Anti-UCHL1 Rabbit Polyclonal Antibody (Biotin)
ABNOPAB14971
: Abnova
- Antigen name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen symbol:UCHL1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Primates,Pig,Horse,Human,Bovine,Guinea pig,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7345
- Antigen synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Amino acid number:100 to 200
- Storage buffer:PBS
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 100-200 of human UCHL1.
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of UCHL1.
Recommended Dilutions: Immunohistochemistry: 2.5-5 ?g/ml; Western Blot: 0.5-2 ?g/ml
Type:
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Guinea Pig, Horse, Human, Mouse, Pig, Primates, Rat