Anti-COL4A4 Rabbit Polyclonal Antibody
ABNOPAB17512
: Abnova
- Antibody type:Primary
- Antigen name:Collagen, type IV, alpha 4
- Antigen symbol:COL4A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:1286
- Antigen synonyms:CA44
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human COL4A4.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of COL4A4.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:1000
Type: Primary
Antigen: COL4A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human