Order Entry
Netherlands
ContactUsLinkComponent
Anti-COL4A3BP Rabbit Polyclonal Antibody
Anti-COL4A3BP Rabbit Polyclonal Antibody
Catalog # ABNOPAB9837
Supplier:  Abnova
Anti-COL4A3BP Rabbit Polyclonal Antibody
Catalog # ABNOPAB9837
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Collagen, type IV, alpha 3 (Goodpasture antigen) binding protein
  • Antigen symbol:
    COL4A3BP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    10087
  • Antigen synonyms:
    STARD11|CERT|CERTL|GPBP|MRD34
  • Storage buffer:
    PBS (0,08% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human COL4A3BP.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of COL4A3BP.

Recommended Dilutions: Western Blot: 5-10 ?g/ml

Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human