Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-NK1.1 Mouse Antibody (redFluor™ 710) [clone: PK136]
 
undefined
Anti-NK1.1 Mouse Antibody (redFluor™ 710) [clone: PK136]

 

  • Catalog No:
  • 80-5941-U100
  • 80-5941-U025
  • Antigen symbol:
  • KLRB1C
  • KLRB1C
  • Antigen name:
  • Killer Cell Lectin-Like Receptor Subfamily B, Member 1
  • Killer Cell Lectin-Like Receptor Subfamily B, Member 1
  • Antigen synonyms:
  • NK-RP1|ly-55c|Nk1|AI462337|NKR-P1.9|Ly-59|Nk-1.2|NK1.1|Ly55c|Ly59|Nk-1|Nkrp1c|NKRP140|CD161|Nk1.2|NKRP1
  • NK-RP1|ly-55c|Nk1|AI462337|NKR-P1.9|Ly-59|Nk-1.2|NK1.1|Ly55c|Ly59|Nk-1|Nkrp1c|NKRP140|CD161|Nk1.2|NKRP1
  • Conjugation:
  • redFluor® 710
  • redFluor® 710
  • Clone:
  • PK136
  • PK136
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 17059
  • 17059
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Store Undiluted at 2 to 8 °C
  • Store Undiluted at 2 to 8 °C

 

Frequently Bought Together