Order Entry
Netherlands
ContactUsLinkComponent
Anti-Zrp Rabbit Polyclonal Antibody [clone: O-146]
Anti-Zrp Rabbit Polyclonal Antibody [clone: O-146]
Supplier:  Cayman Chemical
Certificates
Anti-Zrp Rabbit Polyclonal Antibody [clone: O-146]
Supplier:  Cayman Chemical

Specifications

  • Catalog No:
  • CAYM171142-500
  • CAYM171142-100
  • Antigen symbol:
  • Zrp
  • Zrp
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • O-146
  • O-146
  • Reactivity:
  • Arctic char, Atlantic cod, Rainbow trout, Flounder, Halibut, Turbot, Brown trout, Atlantic salmon
  • Arctic char, Atlantic cod, Rainbow trout, Flounder, Halibut, Turbot, Brown trout, Atlantic salmon
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • eggshell proteins (zona radiata and small amounts of zona pellucida proteins) from Atlantic salmon (S. salar)
  • eggshell proteins (zona radiata and small amounts of zona pellucida proteins) from Atlantic salmon (S. salar)
  • Purification:
  • purified
  • purified
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
  • 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
  • Storage temperature:
  • +4 °C
  • +4 °C
  • Shipping temperature:
  • 4 °C
  • 4 °C

Specifications

Frequently Bought Together