Anti-CACNA1C Mouse Monoclonal Antibody (Unconjugated) [clone: S57-46]
Catalog # ROCK200-301-F08
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Calcium Channel, Voltage-dependent, L type, alpha 1C subunit
- Antigen symbol:CACNA1C
- Clonality:Monoclonal
- Clone:S57-46
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:100144322
- Antigen synonyms:cardic dihydropyridine-sensitive|alpha-1 subunit|voltage-gated calcium channel alpha subunit Cav1.2|calcium channel|isoform 1|voltage-gated L-type calcium channel Cav1.2 alpha 1 subunit|L type|TS|MGC120730|CACN2|CACNL1A1|CCHL1A1|alpha 1C subunit|CACNA1C|cardic muscle|alpha 1 polypeptide|voltage-dependent L-type calcium channel alpha 1C subunit|splice variant 10*|DHPR|voltage-dependent|CaV1.2|CACH2
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:Cav1.2 Antibody was produced in mice by repeated immunizations with a fusion Protein (intracellular carboxyl terminus) of rabbit Cav1.2.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-CAV1.2 Antibody is suitable for use in WB, IP and IHC. Expect a band approximately ~240kDa (varies with cell background due to glycosylation) on specific lysates. Specific conditions for reactivity should be optimised by the end user.
Type: Primary
Antigen: CACNA1C
Clonality: Monoclonal
Clone: S57-46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Rabbit