Anti-STAT5A Mouse Monoclonal Antibody (Unconjugated) [clone: 5F6.F1]
Catalog # ROCK200-301-A45
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Signal transducer and activator of transcription 5A
- Antigen symbol:STAT5A
- Clonality:Monoclonal
- Clone:5F6.F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:20850
- Antigen synonyms:AA959963|STAT5
- Modification:phospho-Tyr694
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-STAT5 phospho Y694 monoclonal antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y694 of mouse STAT5a Protein.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Phospho STAT5 pY694 monoclonal antibody is suitable for Immunofluorescence microscopy, ELISA, immunochemistry and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 91 kDa in size.
Recommended Dilutions: ELISA: 1:20000; Immunofluorescence: 1:50-1:1000; Western Blot: 1:500 - 1:2000; Immunochemistry: 20-40 µg/ml;
Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 5F6.F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Mouse