Anti-ADGRE5 Rabbit Polyclonal Antibody (Unconjugated) [clone: 7C10D8]
Catalog # ROCK200-401-908
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:adhesion G protein-coupled receptor E5
- Antigen symbol:ADGRE5
- Clonality:Polyclonal
- Clone:10H11.E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Chimpanzee,Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:26364
- Antigen synonyms:TM7LN1|AA409984|Cd97
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant Protein corresponding to amino acids 1-512 (extracellular domain) of mouse CD97 Protein.
- Purification:Protein A purified
- Size:500 µg
- Pk:500 µG
Specifications
About this item
This Protein A purified antibody has been tested for use in immunofluorescence microscopy and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 81 kDa in size.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Western Blot: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: ADGRE5
Clonality: Polyclonal
Clone: 7C10D8
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee, Mouse