Order Entry
Netherlands
ContactUsLinkComponent
Anti-TAG-72 Mouse Monoclonal Antibody [clone: B72.3]
Anti-TAG-72 Mouse Monoclonal Antibody [clone: B72.3]
Supplier:  US Biological
Certificates
Anti-TAG-72 Mouse Monoclonal Antibody [clone: B72.3]
Supplier:  US Biological

Specifications

  • Catalog No:
  • USBI138595-100
  • USBI138595
  • Antigen symbol:
  • TAG-72
  • TAG-72
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • B72.3
  • B72.3
  • Reactivity:
  • Hamster, Human, Bovine, Dog, Rat
  • Hamster, Human, Bovine, Dog, Rat
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Membrane-enriched fraction of a human breast carcinoma liver metastasis
  • Membrane-enriched fraction of a human breast carcinoma liver metastasis
  • Form:
  • liquid
  • liquid
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Concentration:
  • ~0,2 mg/ml
  • ~0,2 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, 0,05% BSA, 0,05% sodium azide.
  • PBS, 0,05% BSA, 0,05% sodium azide.
  • Storage temperature:
  • –20 °C
  • –20 °C

Specifications

Related Information

Frequently Bought Together