Anti-Neuropeptide FF Rabbit Polyclonal Antibody
Supplier: US Biological
Certificates
About this item
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Neuropeptide FF
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Specifications
- Catalog No:
- USBI141627-100
- USBI141627
- Antigen name:
- Neuropeptide FF
- Neuropeptide FF
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- liquid
- liquid
- Purification:
- purified by immunoaffinity chromatography
- purified by immunoaffinity chromatography
- Concentration:
- 200 ug/ml
- 200 ug/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- PBS, pH 7,2, 0,05% sodium azide, 50% glycerol.
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together