Anti-Parathyroid Hormone Mouse Monoclonal Antibody [clone: 77/78]
Supplier: US Biological
Certificates
About this item
Recommended dilution: ELISA: 50 ng/ml; Immunochemistry: 1 ?g/ml
Type: Primary
Antigen: Parathyroid Hormone
Clonality: Monoclonal
Clone: 77/78
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,
Specifications
- Catalog No:
- USBIP3109-07A-10
- USBIP3109-07A-100
- Antigen name:
- Parathyroid Hormone
- Parathyroid Hormone
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 77/78
- 77/78
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic human PTH (aa 1-37) polylysine conjugated
- Synthetic human PTH (aa 1-37) polylysine conjugated
- Amino acid number:
- 1 to 37
- 1 to 37
- Form:
- lyophilized powder
- lyophilized powder
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- 50mM Tris, pH 7,2, 0,05% thimerosal.
- 50mM Tris, pH 7,2, 0,05% thimerosal.
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together