Anti-Glutathione S-Transferase pi Mouse Monoclonal Antibody [clone: D2-62]
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Glutathione S-Transferase pi
Clonality: Monoclonal
Clone: D2-62
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human,
- Catalog No:
- USBIG8135-37A-100
- USBIG8135-37A-10
- Antigen symbol:
- GSTP1
- GSTP1
- Antigen name:
- Glutathione S-transferase pi 1
- Glutathione S-transferase pi 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- D2-62
- D2-62
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Human Glutathion S-Transferase p
- Human Glutathion S-Transferase p
- Form:
- lyophilized powder
- lyophilized powder
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- 50mM Tris, pH 7,2.
- 50mM Tris, pH 7,2.
- Storage temperature:
- –20 °C
- –20 °C
- Tested applications:
- Radio Immunoassay
- Radio Immunoassay
Frequently Bought Together