Order Entry
Netherlands
ContactUsLinkComponent
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 3-21]
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 3-21]
Supplier:  US Biological
Certificates
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 3-21]
Supplier:  US Biological

Specifications

  • Catalog No:
  • USBIB2702-33F-10
  • USBIB2702-33F-100
  • Antigen symbol:
  • BNP
  • BNP
  • Antigen name:
  • Brain Natriuretic Peptide
  • Brain Natriuretic Peptide
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 3-21
  • 3-21
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2b
  • IgG2b
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic human BNP (aa 1-32) KLH conjugated (SPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH)
  • Synthetic human BNP (aa 1-32) KLH conjugated (SPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH)
  • Amino acid number:
  • 1 to 32
  • 1 to 32
  • Sequence:
  • SPKMVQGSGCFGRKMDRISSSSGLGCKVLRRH
  • Form:
  • lyophilized powder
  • lyophilized powder
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Concentration:
  • ~1 mg/ml
  • ~1 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,2
  • PBS, pH 7,2
  • Storage temperature:
  • –20 °C
  • –20 °C

Specifications

Related Information

Frequently Bought Together