Anti-Methionine Adenosyltransferase II, alpha Mouse Monoclonal Antibody [clone: 9j18]
USBIM3015-61B-100
- Antibody type:Primary
- Antigen name:Methionine Adenosyltransferase 2 alpha
- Antigen symbol:MAT2A
- Clonality:Monoclonal
- Clone:9j18
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Amino acid number:1 to 395
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Recombinant protein corresponding to aa1-395 of human MAT2A expressed in E. coli (NP_005902).
- Purification:purified by Protein G chromatography
- Size:100 μl
- Pk:100 µl
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: Methionine Adenosyltransferase II, alpha
Clonality: Monoclonal
Clone: 9j18
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human,