Anti-Amyloid 1-40, 42 Mouse Monoclonal Antibody (Biotin) [clone: 6D4]
USBIA2275-75L1
- Antibody type:Primary
- Antigen name:Amyloid 1-40, 42
- Clonality:Monoclonal
- Clone:6D4
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Amino acid number:1 to 42
- Storage buffer:PBS, 0,09% sodium azide, before the addition of glycerol to 40%
- Storage temperature:–20 °C
- Concentration:~0,25 mg/ml
- Immunogen:Synthetic peptide corresponding to aa 1-40 and aa 1-42 of C-terminal human b-amyloid (KLH).
- Purification:purified by thiophilic adsorption and size exclusion chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: Western Blot: 1 ?g/ml; ELISA: 0. 05 ?g/ml; Immunochemistry: 1-10 ?g/ml
Type: Primary
Antigen: Amyloid 1-40, 42
Clonality: Monoclonal
Clone: 6D4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,